Assay Details
| Gene Symbol: TMEM14A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266637.1 | NP_001253566.1 | 147 | 71 | ||
| GenBank mRNA | JU334141.1 | - | 121 | 71 | ||
| JV044100.1 | - | 147 | 71 | |||
| JV639287.1 | - | 161 | 71 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001266637.1 transcripts.
Set Membership: |
|

TMEM14A
Genomic Map