Assay Details
| Gene Symbol: PSMB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265704.1 | NP_001252633.1 | 515 | 62 | ||
| GenBank mRNA | JU327256.1 | - | 448 | 62 | ||
| JU489920.1 | - | 473 | 62 | |||
| JV047701.1 | - | 522 | 62 | |||
| JV656393.1 | - | 165 | 62 | |||
| JV667430.1 | - | 515 | 62 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PSMB4
Genomic Map