Assay Details
| Gene Symbol: AP5S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194748.1 | NP_001181677.1 | 429 | 60 | ||
| XM_015149381.2 | XP_015004867.1 | 274 | 60 | |||
| XM_015149382.2 | XP_015004868.1 | 490 | 60 | |||
| XM_015149383.2 | XP_015004869.1 | 526 | 60 | |||
| GenBank mRNA | JU335318.1 | - | 394 | 60 | ||
| JU476272.1 | - | 388 | 60 | |||
| JV047557.1 | - | 402 | 60 | |||
More Information
Set Membership: |
|

AP5S1
Genomic Map