Assay Details
| Gene Symbol: NT5C3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261493.1 | NP_001248422.1 | 780 | 130 | ||
| XM_015133735.2 | XP_014989221.1 | 1299 | 130 | |||
| XM_015133736.2 | XP_014989222.1 | 1155 | 130 | |||
| GenBank mRNA | JU334836.1 | - | 732 | 130 | ||
| JU472937.1 | - | 780 | 130 | |||
More Information
Set Membership: |
|

NT5C3A
Genomic Map