Assay Details
| Gene Symbol: C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136030.2 | XP_014991516.1 | 1626 | 86 | ||
| XM_015136031.2 | XP_014991517.1 | 617 | 86 | |||
| GenBank mRNA | JU326890.1 | - | 855 | 86 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

C2
Genomic Map