Assay Details
| Gene Symbol: PTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194354.2 | NP_001181283.1 | 87 | 80 | ||
| XM_015134931.2 | XP_014990417.1 | 553 | 80 | |||
| XM_015134933.2 | XP_014990419.1 | 553 | 80 | |||
| GenBank mRNA | JU321403.1 | - | 371 | 80 | ||
| JV045303.1 | - | 217 | 80 | |||
| JV588958.1 | - | 354 | 80 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194354.2, XM_015134931.2, XM_015134933.2 transcripts.
Set Membership: |
|

PTN
Genomic Map