Assay Details
| Gene Symbol: SLC25A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266706.1 | NP_001253635.1 | 709 | 76 | ||
| XM_028827444.1 | XP_028683277.1 | 403 | 76 | |||
| GenBank mRNA | JU336010.1 | - | 636 | 76 | ||
| JU472268.1 | - | 709 | 76 | |||
| JV659371.1 | - | 742 | 76 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SLC25A1
Genomic Map