Assay Details
| Gene Symbol: PPIB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266312.1 | NP_001253241.1 | 210 | 83 | ||
| GenBank mRNA | JU328179.1 | - | 101 | 83 | ||
| JU471031.1 | - | 210 | 83 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PPIB
Genomic Map