Assay Details
| Gene Symbol: LTB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047150.1 | NP_001040615.1 | 282 | 109 | ||
| XM_015135928.2 | XP_014991414.1 | 260 | 109 | |||
| GenBank mRNA | JU486647.1 | - | 149 | 109 | ||
More Information
Set Membership: |
|

LTB
Genomic Map