Assay Details
| Gene Symbol: PSMB9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194864.2 | NP_001181793.1 | 167 | 58 | ||
| NM_001376986.1 | NP_001363915.1 | 205 | 58 | |||
| XM_015136060.2 | XP_014991546.1 | 600 | 58 | |||
| GenBank mRNA | JU322675.1 | - | 205 | 58 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PSMB9
Genomic Map