Assay Details
| Gene Symbol: PRR13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266483.1 | NP_001253412.1 | 97 | 65 | ||
| GenBank mRNA | JU335071.1 | - | 98 | 65 | ||
| JV047427.1 | - | 97 | 65 | |||
| JV654146.1 | - | 113 | 65 | |||
| JV654147.1 | - | 113 | 65 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001266483.1 transcripts.
Set Membership: |
|

PRR13
Genomic Map