Assay Details
| Gene Symbol: RGS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258139.1 | NP_001245068.1 | 445 | 130 | ||
| GenBank mRNA | JU325546.1 | - | 153 | 130 | ||
| JU476668.1 | - | 445 | 130 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RGS1
Genomic Map