Assay Details
| Gene Symbol: DDAH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260633.2 | NP_001247562.1 | 552 | 83 | ||
| XM_015135937.2 | XP_014991423.1 | 963 | 83 | |||
| XM_015135939.2 | XP_014991425.1 | 642 | 83 | |||
| XM_015135940.2 | XP_014991426.1 | 489 | 83 | |||
| GenBank mRNA | JU327342.1 | - | 288 | 83 | ||
| JV045619.1 | - | 554 | 83 | |||
| JV500946.1 | - | 573 | 83 | |||
| JV602516.1 | - | 566 | 83 | |||
| JV635547.1 | - | 554 | 83 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DDAH2
Genomic Map