Assay Details
| Gene Symbol: POLD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266109.1 | NP_001253038.1 | 311 | 82 | ||
| GenBank mRNA | JU322440.1 | - | 372 | 82 | ||
| JV044500.1 | - | 311 | 82 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

POLD4
Genomic Map