Assay Details
| Gene Symbol: GJA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261067.1 | NP_001247996.1 | 161 | 63 | ||
| XM_015137308.2 | XP_014992794.1 | 254 | 63 | |||
| GenBank mRNA | JU322239.1 | - | 215 | 63 | ||
| JU472826.1 | - | 169 | 63 | |||
| JV046753.1 | - | 161 | 63 | |||
| JV565908.1 | - | 192 | 63 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001261067.1, XM_015137308.2 transcripts.
Set Membership: |
|

GJA1
Genomic Map