Assay Details
| Gene Symbol: RBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015132011.2 | XP_014987497.1 | 563 | 70 | ||
| GenBank mRNA | JU346727.1 | - | 529 | 70 | ||
| JV657891.1 | - | 541 | 70 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RBP1
Genomic Map