Assay Details
| Gene Symbol: HDGFL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015142890.2 | XP_014998376.1 | 609 | 126 | ||
| XM_015142891.2 | XP_014998377.1 | 1032 | 126 | |||
| GenBank mRNA | JU334742.1 | - | 612 | 126 | ||
| JV047218.1 | - | 565 | 126 | |||
| JV667957.1 | - | 610 | 126 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HDGFL3
Genomic Map