Assay Details
| Gene Symbol: C1H1orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015110674.2 | XP_014966160.1 | 955 | 133 | ||
| XM_015110681.2 | XP_014966167.1 | 901 | 133 | |||
| XM_015110691.2 | XP_014966177.1 | 878 | 133 | |||
| XM_028828709.1 | XP_028684542.1 | 586 | 133 | |||
| XM_028828715.1 | XP_028684548.1 | 728 | 133 | |||
| GenBank mRNA | JU319736.1 | - | 841 | 133 | ||
| JU333869.1 | - | 787 | 133 | |||
| JU471218.1 | - | 824 | 133 | |||
| JV046682.1 | - | 775 | 133 | |||
| JV636653.1 | - | 894 | 133 | |||
| JV711518.1 | - | 784 | 133 | |||
More Information
Set Membership: |
|

C1H1orf43
Genomic Map