Assay Details
| Gene Symbol: CFAP410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015132771.2 | XP_014988257.2 | 578 | 78 | ||
| XM_015132772.2 | XP_014988258.2 | 578 | 78 | |||
| XM_028845283.1 | XP_028701116.1 | 606 | 78 | |||
| XM_028845284.1 | XP_028701117.1 | 607 | 78 | |||
| XM_028845285.1 | XP_028701118.1 | 2234 | 78 | |||
| XM_028845286.1 | XP_028701119.1 | 2234 | 78 | |||
| GenBank mRNA | JU336275.1 | - | 563 | 78 | ||
| JU477114.1 | - | 463 | 78 | |||
| JV048182.1 | - | 405 | 78 | |||
| JV660663.1 | - | 585 | 78 | |||
| JV667472.1 | - | 452 | 78 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CFAP410
Genomic Map