Assay Details
| Gene Symbol: BUD23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266959.1 | NP_001253888.1 | 72 | 56 | ||
| GenBank mRNA | JU332790.1 | - | 78 | 56 | ||
| JU332791.1 | - | 78 | 56 | |||
| JU332792.1 | - | 78 | 56 | |||
| JU474210.1 | - | 78 | 56 | |||
| JV043191.1 | - | 72 | 56 | |||
| JV706194.1 | - | 94 | 56 | |||
More Information
Set Membership: |
|

BUD23
Genomic Map