Assay Details
| Gene Symbol: PLPP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015140445.2 | XP_014995931.1 | 1022 | 121 | ||
| GenBank mRNA | JU336872.1 | - | 768 | 121 | ||
| JU477610.1 | - | 402 | 121 | |||
| JV045043.1 | - | 789 | 121 | |||
| JV045044.1 | - | 786 | 121 | |||
| JV561387.1 | - | 741 | 121 | |||
| JV602210.1 | - | 845 | 121 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PLPP3
Genomic Map