Assay Details
| Gene Symbol: APCS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001117234.4 | XP_001117234.2 | 164 | 148 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001117234.4 transcripts.
Set Membership: |
|

APCS
Genomic Map