Assay Details
| Gene Symbol: FCGRT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028839693.1 | XP_028695526.1 | 801 | 105 | ||
| XM_028839694.1 | XP_028695527.1 | 774 | 105 | |||
| XM_028839695.1 | XP_028695528.1 | 769 | 105 | |||
| XM_028839696.1 | XP_028695529.1 | 745 | 105 | |||
| XM_028839697.1 | XP_028695530.1 | 775 | 105 | |||
| XM_028839698.1 | XP_028695531.1 | 821 | 105 | |||
| XM_028839699.1 | XP_028695532.1 | 800 | 105 | |||
| XM_028839700.1 | XP_028695533.1 | 762 | 105 | |||
| Gene Symbol: LOC718859 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257520.1 | NP_001244449.1 | 481 | 105 | ||
| GenBank mRNA | JU322969.1 | - | 481 | 105 | ||
| JU474694.1 | - | 561 | 105 | |||
| JV044676.1 | - | 667 | 105 | |||
| JV044677.1 | - | 561 | 105 | |||
More Information
Set Membership: |
|

FCGRT
Genomic Map