Assay Details
| Gene Symbol: C1H1orf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260863.1 | NP_001247792.1 | 518 | 122 | ||
| XM_028845165.1 | XP_028700998.1 | 422 | 122 | |||
| GenBank mRNA | JU319728.1 | - | 741 | 122 | ||
| JU477354.1 | - | 555 | 122 | |||
| JV048336.1 | - | 518 | 122 | |||
| JV566003.1 | - | 768 | 122 | |||
| JV601878.1 | - | 801 | 122 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

C1H1orf21
Genomic Map