Assay Details
| Gene Symbol: MAMU-DMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193474.1 | NP_001180403.1 | 426 | 136 | ||
| GenBank mRNA | CO725887.1 | - | 429 | 136 | ||
| CO726452.1 | - | 382 | 136 | |||
| JU325668.1 | - | 121 | 136 | |||
| JV049020.1 | - | 418 | 136 | |||
More Information
Set Membership: |
|

MAMU-DMA
Genomic Map