Assay Details
| Gene Symbol: TMEM222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015132597.2 | XP_014988083.1 | 736 | 58 | ||
| XM_015132606.2 | XP_014988092.1 | 640 | 58 | |||
| GenBank mRNA | JU326119.1 | - | 533 | 58 | ||
| JU477451.1 | - | 544 | 58 | |||
More Information
Set Membership: |
|

TMEM222
Genomic Map