Assay Details
| Gene Symbol: DPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118202.2 | XP_014973688.1 | 986 | 76 | ||
| XM_028835865.1 | XP_028691698.1 | 941 | 76 | |||
| XM_028835866.1 | XP_028691699.1 | 1019 | 76 | |||
| XM_028835867.1 | XP_028691700.1 | 1125 | 76 | |||
| GenBank mRNA | JU325578.1 | - | 912 | 76 | ||
| JV535594.1 | - | 866 | 76 | |||
More Information
Set Membership: |
|

DPH1
Genomic Map