Assay Details
| Gene Symbol: RPL30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001094073.4 | XP_001094073.1 | 147 | 151 | ||
| XM_028852790.1 | XP_028708623.1 | 146 | 151 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001094073.4, XM_028852790.1 transcripts.
Set Membership: |
|

RPL30
Genomic Map