Assay Details
| Gene Symbol: RPL30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001094073.4 | XP_001094073.1 | 420 | 131 | ||
| XM_028852790.1 | XP_028708623.1 | 419 | 131 | |||
| GenBank mRNA | JU491121.1 | - | 291 | 131 | ||
| JV692600.1 | - | 272 | 131 | |||
More Information
Set Membership: |
|

RPL30
Genomic Map