Assay Details
| Gene Symbol: TFG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257800.1 | NP_001244729.1 | 1111 | 158 | ||
| XM_015128711.2 | XP_014984197.1 | 1007 | 158 | |||
| XM_015128715.2 | XP_014984201.1 | 1027 | 158 | |||
| XM_015128719.2 | XP_014984205.1 | 1213 | 158 | |||
| XM_028843166.1 | XP_028698999.1 | 882 | 158 | |||
| GenBank mRNA | JU319634.1 | - | 945 | 158 | ||
| JU319635.1 | - | 1111 | 158 | |||
| JU470541.1 | - | 1120 | 158 | |||
| JV043772.1 | - | 1087 | 158 | |||
| JV703431.1 | - | 946 | 158 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TFG
Genomic Map