Assay Details
| Gene Symbol: GSTM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261651.1 | NP_001248580.1 | 428 | 147 | ||
| GenBank mRNA | JU475302.1 | - | 428 | 147 | ||
| JV542578.1 | - | 321 | 147 | |||
| JV579670.1 | - | 664 | 147 | |||
| KC734498.1 | - | 359 | 147 | |||
More Information
Set Membership: |
|

GSTM2
Genomic Map