Assay Details
| Gene Symbol: PDLIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001099155.4 | XP_001099155.1 | 951 | 146 | ||
| GenBank mRNA | JU477770.1 | - | 624 | 146 | ||
| JV048647.1 | - | 598 | 146 | |||
| JV636445.1 | - | 627 | 146 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PDLIM1
Genomic Map