Assay Details
| Gene Symbol: PEBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246199.1 | NP_001233128.1 | 242 | 152 | ||
| GenBank mRNA | JF693320.1 | - | 242 | 152 | ||
| JU475772.1 | - | 306 | 152 | |||
| JV047201.1 | - | 1244 | 152 | |||
| JV047202.1 | - | 1244 | 152 | |||
| JV047203.1 | - | 1244 | 152 | |||
| JV047204.1 | - | 1244 | 152 | |||
| JV047205.1 | - | 1244 | 152 | |||
| JV047206.1 | - | 1244 | 152 | |||
| JV047207.1 | - | 1244 | 152 | |||
| JV047208.1 | - | 1244 | 152 | |||
| JV635384.1 | - | 390 | 152 | |||
| JV636485.1 | - | 390 | 152 | |||
More Information
Set Membership: |
|

PEBP1
Genomic Map