Assay Details
| Gene Symbol: GOT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126312.2 | XP_014981798.2 | 721 | 117 | ||
| XM_028841324.1 | XP_028697157.1 | 721 | 117 | |||
| GenBank mRNA | JU334069.1 | - | 703 | 117 | ||
| JU475231.1 | - | 677 | 117 | |||
| JV046797.1 | - | 716 | 117 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GOT2
Genomic Map