Assay Details
| Gene Symbol: RPS19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124097.1 | XP_014979583.1 | 385 | 68 | ||
| GenBank mRNA | JU491192.1 | - | 87 | 68 | ||
| JV658228.1 | - | 116 | 68 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015124097.1 transcripts.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RPS19
Genomic Map