Assay Details
| Gene Symbol: ACO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261164.2 | NP_001248093.1 | 1326 | 119 | ||
| XM_015150398.2 | XP_015005884.1 | 1266 | 119 | |||
| XM_028827304.1 | XP_028683137.1 | 1570 | 119 | |||
| GenBank mRNA | JV047194.1 | - | 1327 | 119 | ||
| JV583174.1 | - | 1316 | 119 | |||
| JV636183.1 | - | 1326 | 119 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ACO2
Genomic Map