Assay Details
| Gene Symbol: POP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261725.2 | NP_001248654.1 | 226 | 85 | ||
| GenBank mRNA | JU321174.1 | - | 67 | 85 | ||
| JU472610.1 | - | 230 | 85 | |||
| JV603184.1 | - | 249 | 85 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001261725.2 transcripts.
Set Membership: |
|

POP7
Genomic Map