Assay Details
| Gene Symbol: PPP1R11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047136.1 | NP_001040601.1 | 175 | 147 | ||
| XM_015135852.2 | XP_014991338.1 | 237 | 147 | |||
| XM_028846796.1 | XP_028702629.1 | 996 | 147 | |||
| GenBank mRNA | JU320683.1 | - | 372 | 147 | ||
| JU320684.1 | - | 372 | 147 | |||
| JU472485.1 | - | 189 | 147 | |||
| JV045668.1 | - | 333 | 147 | |||
| JV566886.1 | - | 380 | 147 | |||
| JV703696.1 | - | 315 | 147 | |||
More Information
Set Membership: |
|

PPP1R11
Genomic Map