Assay Details
| Gene Symbol: NLRP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114351.1 | NP_001107823.1 | 3006 | 74 | ||
| XM_015127781.2 | XP_014983267.2 | 4135 | 74 | |||
| GenBank mRNA | EU344966.1 | - | 3006 | 74 | ||
More Information
Set Membership: |
|

NLRP3
Genomic Map