Assay Details
| Gene Symbol: MRGBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148633.2 | XP_015004119.1 | 337 | 69 | ||
| GenBank mRNA | JU328209.1 | - | 120 | 69 | ||
| JV047536.1 | - | 281 | 69 | |||
| JV655248.1 | - | 318 | 69 | |||
More Information
Set Membership: |
|

MRGBP
Genomic Map