Assay Details
Gene Symbol: CCNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015142387.2 | XP_014997873.1 | 1084 | 110 | ||
GenBank mRNA | JU473590.1 | - | 576 | 110 | ||
JV045721.1 | - | 537 | 110 | |||
JV635466.1 | - | 565 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
