Assay Details
| Gene Symbol: PMM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150464.2 | XP_015005950.1 | 458 | 81 | ||
| XM_015150465.2 | XP_015005951.1 | 499 | 81 | |||
| GenBank mRNA | JU335037.1 | - | 401 | 81 | ||
| JU476036.1 | - | 383 | 81 | |||
| JV619594.1 | - | 439 | 81 | |||
More Information
Set Membership: |
|

PMM1
Genomic Map