Assay Details
| Gene Symbol: RAN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260776.1 | NP_001247705.1 | 366 | 117 | ||
| XM_015109692.2 | XP_014965178.1 | 416 | 117 | |||
| GenBank mRNA | JU327157.1 | - | 301 | 117 | ||
| JU476635.1 | - | 392 | 117 | |||
| JV047818.1 | - | 366 | 117 | |||
More Information
Set Membership: |
|

RAN
Genomic Map