Assay Details
Gene Symbol: KHDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257908.1 | NP_001244837.1 | 646 | 128 | ||
XM_015136730.2 | XP_014992216.2 | 690 | 128 | |||
XM_015136731.2 | XP_014992217.1 | 631 | 128 | |||
XM_015136732.2 | XP_014992218.1 | 1041 | 128 | |||
GenBank mRNA | JU322420.1 | - | 646 | 128 | ||
JV044900.1 | - | 598 | 128 | |||
JV594694.1 | - | 673 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
