Assay Details
| Gene Symbol: COA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193378.1 | NP_001180307.1 | 256 | 131 | ||
| GenBank mRNA | DR767791.1 | - | 260 | 131 | ||
| JU334845.1 | - | 273 | 131 | |||
| JU475873.1 | - | 282 | 131 | |||
| JV047275.1 | - | 239 | 131 | |||
More Information
Set Membership: |
|

COA4
Genomic Map