Assay Details
| Gene Symbol: AMD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015137572.2 | XP_014993058.1 | 1416 | 109 | ||
| GenBank mRNA | JU322126.1 | - | 1383 | 109 | ||
| JU322127.1 | - | 1383 | 109 | |||
| JU471108.1 | - | 1380 | 109 | |||
| JU475120.1 | - | 1359 | 109 | |||
| JV043597.1 | - | 1309 | 109 | |||
| JV451998.1 | - | 1384 | 109 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

AMD1
Genomic Map