Assay Details
| Gene Symbol: SP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028830845.1 | XP_028686678.1 | 3261 | 163 | ||
| XM_028830846.1 | XP_028686679.1 | 3219 | 163 | |||
| GenBank mRNA | JU492091.1 | - | 2669 | 163 | ||
| JV659586.1 | - | 2672 | 163 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SP3
Genomic Map