Assay Details
| Gene Symbol: YBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015137147.2 | XP_014992633.1 | 940 | 74 | ||
| XM_015137151.2 | XP_014992637.1 | 850 | 74 | |||
| GenBank mRNA | JU328105.1 | - | 471 | 74 | ||
| JU487384.1 | - | 701 | 74 | |||
| JU487387.1 | - | 701 | 74 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

YBX1
Genomic Map