Assay Details
| Gene Symbol: BOLA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150500.2 | XP_015005986.1 | 761 | 99 | ||
| XM_028842221.1 | XP_028698054.1 | 951 | 99 | |||
| GenBank mRNA | JU473818.1 | - | 770 | 99 | ||
| JV045581.1 | - | 699 | 99 | |||
| JV685698.1 | - | 390 | 99 | |||
More Information
Set Membership: |
|

BOLA1
Genomic Map