Assay Details
| Gene Symbol: NIP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126493.2 | XP_014981979.1 | 1147 | 163 | ||
| GenBank mRNA | JV639306.1 | - | 933 | 163 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NIP7| Gene Symbol: NIP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126493.2 | XP_014981979.1 | 1147 | 163 | ||
| GenBank mRNA | JV639306.1 | - | 933 | 163 | ||
Set Membership: |
|